Learn More
Sartorius Polyethersulfone Membrane Filters
Polyethersulfone (PES) filters have a high internal porosity.
Brand:  Sartorius 15407--25----MIN
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11733693
  • 1316.00 SEK / Pack of 100
Estimated Shipment
20-04-2021
Add to basket
Description
Description
Polyethersulfone (PES) filters have a high internal porosity.
They perform well at high flux with an excellent throughput of aqueous solutions over the entire pH range of 1-14. Thanks to their low non-specific protein adsorption the PES type is recommended for filtering biological and pharmaceutical solutions.
Specifications
Specifications
0.2μm
25mm
Recommended for filtering biological and pharmaceutical solutions.
Polyethersulfone (PES)
100/pk.
150μm