missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Quantitative Grade 389 F Filter Papers
Brand: Sartorius FT-3-112-110
Additional Details : Weight : 0.01000kg
Description
- Ash-free (ash content ≤ 0.01 % according to DIN 540370)
- Wet-strengthened
- Color-coded box for easy selection
Specifications
20 sec. | |
100 per pack | |
Disc | |
Yellow |
8 to 12μmm | |
389 | |
110mm | |
0.19mm |