Learn More
Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Product Code. 11708599 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Acid Neutralization Cartridge
Acid Neutralizing Compound
Ammonia Neutralizing Solution
Cart
Chemical Trap Kit
Condensation Flask
Disposable Cartridge
Heat Transfer Fluid
Tubing Pack
Unit Size:
0.25kg
5 L
Each
Pack of 4
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11708599 Disposable Cartridge 0.25kg
11778629 Chemical Trap Kit Each
11738599 Acid Neutralization Cartridge Each
15297909 Acid Neutralizing Compound Each
11748589 Tubing Pack Each
10399102 Ammonia Neutralizing Solution Each
11748569 Ammonia Neutralizing Solution Pack of 4
12642765 Heat Transfer Fluid 5 L
11728669 Condensation Flask Each
10684283 Cart Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 11708599 Supplier Thermo Scientific™ Supplier No. DC120A
  • ONLINE EXCLUSIVE PRICE
    4677.21 SEK
    3811.07 SEK / 0.25kg
    Save 866.14 SEK
    18% Av
View all promotions
 Restricted Item
Beräknad leverans: 07-12-2026
Improve the functionality of Savant SpeedVac Concentrators with these Savant SpeedVac Concentrators miscellaneous accessories.
Improve the funcionality of Thermo Scientific™ Savant™ SpeedVac™ Concentrators with Savant™ SpeedVac™ Concentrators Accessories.
TRUSTED_SUSTAINABILITY
Specifications
Description Disposable Cartridge (trapping volatile radioactivity)
Type Disposable Cartridge
For Use With (Equipment) All SpeedVac™ Unit
compliance-icons compliance-icons
Product Identifier
  • Cryocool Heat Transfer Fluid
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P370+P378-In case of fire: Use dry sanddry chemicalor alcohol-resistant to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: octamethyltrisiloxane, actamethylcyclote