Learn More
Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Product Code. 11728669 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Acid Neutralization Cartridge
Acid Neutralizing Compound
Ammonia Neutralizing Solution
Cart
Chemical Trap Kit
Condensation Flask
Disposable Cartridge
Heat Transfer Fluid
Tubing Pack
Unit Size:
0.25kg
5 L
Each
Pack of 4
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11728669 Condensation Flask Each
11778629 Chemical Trap Kit Each
11708599 Disposable Cartridge 0.25kg
11738599 Acid Neutralization Cartridge Each
15297909 Acid Neutralizing Compound Each
11748589 Tubing Pack Each
10399102 Ammonia Neutralizing Solution Each
11748569 Ammonia Neutralizing Solution Pack of 4
12642765 Heat Transfer Fluid 5 L
10684283 Cart Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 11728669 Supplier Thermo Scientific™ Supplier No. GCF400
  • ONLINE EXCLUSIVE PRICE
    12051.45 SEK
    11372.74 SEK / Paket med 1
    Save 678.71 SEK
    5% Av
View all promotions
I lager
Improve the functionality of Savant SpeedVac Concentrators with these Savant SpeedVac Concentrators miscellaneous accessories.
Improve the funcionality of Thermo Scientific™ Savant™ SpeedVac™ Concentrators with Savant™ SpeedVac™ Concentrators Accessories.
TRUSTED_SUSTAINABILITY
Specifications
Description Glass condensation flask
Type Condensation Flask
For Use With (Equipment) SPD1030 and SPD2030 DNA SpeedVac™ Concentrator