Läs mer
Thermo Scientific™ Savant™ SpeedVac™ High Capacity Concentrators
Optimize evaporation with the three selectable heating levels of these high capacity concentrators, ideal for aqueous and organic solvents.
Specifikationer
| Avluftningsventil | Automatic |
|---|---|
| Kammarmaterial | PTFE |
| Avgasportens storlek | Standard |
| Håller | 8 Deepwell, 12 Shallow Well Microplates |
| Magnetisk montering | UMA200 |
Beskrivning
Optimize evaporation with Thermo Scientific™ Savant™ SpeedVac™ High Capacity Concentrators. With three selectable heating levels, these concentrators are ideal for aqueous and organic solvents.
Features:- Large rotor selection to accommodate a variety of tube sizes and types
- PTFE coated chamber provides long lasting durability against harsh solvents
- Automatic bleeder valve automatically provides vacuum at the beginning of a run and bleeds chamber to atmospheric pressure at the end of a run
- Radiant cover maximizes evaporation/sublimation rates (optional)
Ordering Information:
Rotors are not included and should be ordered separately.
Specifikationer
| Automatic | |
| Standard | |
| UMA200 | |
| 18 in. | |
| 18 in. | |
| 25 in. | |
| 102 lb. | |
| 50/60 Hz |
| PTFE | |
| 8 Deepwell, 12 Shallow Well Microplates | |
| 3 Presets – Low, Med, High (ambient, 43°C, 65°C) | |
| 450 mm | |
| 460 mm | |
| 620 mm | |
| 46 kg |