Läs mer
Thermo Scientific™ Savant™ SpeedVac™ High Capacity Concentrators
Optimize evaporation with the three selectable heating levels of these high capacity concentrators, ideal for aqueous and organic solvents.
Specifikationer
Avluftningsventil Automatic
Kammarmaterial PTFE
Avgasportens storlek Standard
Håller 8 Deepwell, 12 Shallow Well Microplates
Magnetisk montering UMA200
Se fler specifikationer
Beskrivning
Beskrivning
Optimize evaporation with Thermo Scientific™ Savant™ SpeedVac™ High Capacity Concentrators. With three selectable heating levels, these concentrators are ideal for aqueous and organic solvents.
Features:
  • Large rotor selection to accommodate a variety of tube sizes and types
  • PTFE coated chamber provides long lasting durability against harsh solvents
  • Automatic bleeder valve automatically provides vacuum at the beginning of a run and bleeds chamber to atmospheric pressure at the end of a run
  • Radiant cover maximizes evaporation/sublimation rates (optional)
Ordering Information:
Rotors are not included and should be ordered separately.
Specifikationer
Specifikationer
Automatic
Standard
UMA200
18 in.
18 in.
25 in.
102 lb.
50/60 Hz
PTFE
8 Deepwell, 12 Shallow Well Microplates
3 Presets – Low, Med, High (ambient, 43°C, 65°C)
450 mm
460 mm
620 mm
46 kg
Filmer
SDS
Dokument
Dokument
Produktspecifikationer