Läs mer
Saxagliptin hydrochloride, Tocris Bioscience™
DPPIV/CD26 Inhibitors
Brand: Tocris Bioscience 6507/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Saxagliptin hydrochloride is a high affinity DPP-IV inhibitor (Ki = 0.6 nM). Inhibits plasma DPP-IV (87%). Elicits enhanced glucose clearance in Zuckerfa/fa rats.Specifikationer
| 709031-78-7 | |
| (1S,3S,5S)-2-[(2S)-Amino(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)acetyl]-2-azabicyclo[3.1.0]hexane-3-carbonitrile hydrochloride | |
| 10 mg |