Läs mer
Saxagliptin Hydrochloride, TRC
Produktkod. 30424965
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30424965 250 mg
1 options
Produktkod. 30424965 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS143753250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Saxagliptin Hydrochloride
CAS 709031-78-7
Molekylformel C18 H25 N3 O2 . Cl H
Kvantitet 250 mg
Synonym 2-Azabicyclo[3.1.0]hexane-3-carbonitrile','2-[(2S)-2-amino-2-(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)acetyl]-','hydrochloride (1:1)','(1S,3S,5S)-, 2-Azabicyclo[3.1.0]hexane-3-carbonitrile','2-[(2S)-amino(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)acetyl]-','monohydrochloride','(1S,3S,5S)- (9CI), Saxagliptin hydrochloride
InChI formel InChI=1S/C18H25N3O2.ClH/c19-8-13-2-12-3-14(12)21(13)16(22)15(20)17-4-10-1-11(5-17)7-18(23,6-10)9-17;/h10-15,23H,1-7,9,20H2;1H/t10?,11?,12-,13+,14+,15-,17?,18?;/m1./s1
LEDER Cl.N[C@H](C(=O)N1[C@@H](C[C@@H]2C[C@H]12)C#N)C34CC5CC(CC(O)(C5)C3)C4
IUPAC-namn (1S,3S,5S)-2-[(2S)-2-amino-2-(3-hydroxy-1-adamantyl)acetyl]-2-azabicyclo[3.1.0]hexane-3-carbonitrile;hydrochloride
Molekylvikt (g/mol) 351.87
Formel vikt 351.1714