Läs mer
Saxagliptin O-Beta-D-glucuronide, TRC
Produktkod. 30413919
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30413919 5 mg
1 options
Produktkod. 30413919 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS1435105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Saxagliptin O-beta-D-glucuronide
CAS 1155849-58-3
Molekylformel C24H33N3O8
Kvantitet 5 mg
Synonym (2S,3S,5R,6S)-6-(((1R,3S,5R,7S)-3-((S)-1-Amino-2-((1S,3S,5S)-3-cyano-2-azabicyclo[3.1.0]hexan-2-yl)-2-oxoethyl)adamantan-1-yl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic Acid
InChI formel InChI=1S/C24H33N3O8/c25-8-13-2-12-3-14(12)27(13)20(31)19(26)23-4-10-1-11(5-23)7-24(6-10,9-23)35-22-17(30)15(28)16(29)18(34-22)21(32)33/h10-19,22,28-30H,1-7,9,26H2,(H,32,33)/t10-,11+,12-,13+,14+,15?,16-,17+,18-,19-,22-,23+,24-/m1/s1
LEDER N[C@H](C(=O)N1[C@@H](C[C@@H]2C[C@H]12)C#N)[C@@]34C[C@@H]5C[C@@H](C[C@@](C5)(C3)O[C@H]6O[C@H]([C@H](O)C(O)[C@@H]6O)C(=O)O)C4
IUPAC-namn (2R,3R,5S,6R)-6-[[(5R,7S)-3-[(1S)-1-amino-2-[(1S,3S,5S)-3-cyano-2-azabicyclo[3.1.0]hexan-2-yl]-2-oxoethyl]-1-adamantyl]oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
Molekylvikt (g/mol) 491.53
Formel vikt 491.2268