Läs mer
Tocris Bioscience™ SB 204990
GREENER_CHOICE
ATP citrate lyase (ACLY) inhibitor
Brand:  Tocris Bioscience™ 4962/50
Encompass
Produktkod. 15273117
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
SB 204990 is an ATP citrate lyase (ACLY) inhibitor. Prodrug of SB 201076. Inhibits cholesterol and fatty acid synthesis in a dose-dependent manner in HepG2 cells. Orally active in vivo.
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Certifikat
Kampanjer
Kampanjer