Läs mer
Tocris Bioscience™ SB 222200
GREENER_CHOICE
Produktkod. 30001242
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30001242 10 mg
18084472 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30001242 Leverantör Tocris Bioscience™ Leverantörsnummer 1393/10
Potent, selective non-peptide NK3 antagonist. Brain penetrant
Potent and selective non-peptide NK3 receptor antagonist (Ki values are 4.4, > 100,000 and 250 nM for human NK3, NK1 and NK2 receptors respectively). Antihypertensive in vivo. Brain penetrant.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 174635-69-9
Molekylformel C26H24N2O
Specifikationer
Kemiskt namn eller material SB 222200
Kvantitet 10 mg
Formel vikt 380.48
Renhet >98%
Molekylvikt (g/mol) 380.491