Läs mer
Tocris Bioscience™ SB 334867
Selective non-peptide OX1 antagonist
Brand: Tocris Bioscience™ 1960/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
SB 334867 is a selective non-peptide orexin OX1 receptor antagonist. pKb values are 7.2 and < 5 for inhibition of intracellular Ca2+ release in CHO cells expressing human OX1 and OX2 receptors respectively. Blocks orexin-A induced grooming and feeding following systemic administration in vivo.Specifikationer
| N-(2-Methyl-6-benzoxazolyl)-N'-1,5-naphthyridin-4-yl urea | |
| >99% | |
| Soluble to 100mM with gentle warming in DMSO |
| 792173-99-0 | |
| 50 mg |
Produkttitel
For Research Use Only