Läs mer
SB-431542, MedChemExpress
Produktkod. 16453968
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16453968 10 mg
16474976 100 mg
16400327 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16453968 Leverantör Medchem Express Leverantörsnummer HY10431
MedChemExpress SB-431542 is a potent and selective inhibitor of ALK5/TGF-β type I Receptor with an IC50 value of 94 nM.
SB-431542 is a potent and selective inhibitor of ALK5/TGF-β type I Receptor with an IC50 value of 94 nM.
Specifikationer
Kemiskt namn eller material SB-431542
CAS 301836-41-9
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C22H16N4O3
Kvantitet 10 mg
Löslighetsinformation DMSO : 250 mg/mL (650.38 mM; Need ultrasonic) ∣Ethanol : 11.17 mg/mL (29.06 mM; Need ultrasonic and warming)
LEDER O=C(C1=CC=C(C=C1)C2=NC(C3=CC=C4OCOC4=C3)=C(N2)C5=NC=CC=C5)N
Molekylvikt (g/mol) 384.39
Formel vikt 384.39
Procent renhet 97.36%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Cancer-Kinase/protease