Läs mer
SB-505124, MedChemExpress
Produktkod. 16461887
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16461887 50 mg
16435728 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16461887 Leverantör Medchem Express Leverantörsnummer HY13521
MedChemExpress SB-505124 is a selective inhibitor of TGF-β Receptor type I receptors (ALK4, ALK5, ALK7), with IC50s of 129 nM and 47 nM for ALK4, ALK5, respectively, but it does not inhibit ALK1, 2, 3, or 6.
SB-505124 is a selective inhibitor of TGF-β Receptor type I receptors (ALK4, ALK5, ALK7), with IC50s of 129 nM and 47 nM for ALK4, ALK5, respectively, but it does not inhibit ALK1, 2, 3, or 6.
Specifikationer
Kemiskt namn eller material SB-505124
CAS 694433-59-5
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C20H21N3O2
Kvantitet 50 mg
Löslighetsinformation DMSO : 113.33 mg/mL (337.90 mM; Need ultrasonic)
LEDER CC1=CC=CC(C2=C(C3=CC=C(OCO4)C4=C3)N=C(C(C)(C)C)N2)=N1
Molekylvikt (g/mol) 335.4
Formel vikt 335.4
Procent renhet 99.63%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease