Läs mer
Tocris Bioscience™ SB 505124
GREENER_CHOICE
Produktkod. 18772734
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18772734 50 mg
18702584 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18772734 Leverantör Tocris Bioscience™ Leverantörsnummer 3263/50
Selective inhibitor of TGF-βRI, ALK4 and ALK7
Selective inhibitor of transforming growth factor-β type I receptor (ALK5), ALK4 and ALK7 (IC50 values are 47 and 129 nM for ALK5 and ALK4 respectively). Selectively inhibits signaling from TGF-β and activin; does not inhibit other ALK family members.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 694433-59-5
Molekylformel C20H21N3O2
Specifikationer
Kemiskt namn eller material SB 505124
Kvantitet 50 mg
Formel vikt 335.4
Renhet >98%
Molekylvikt (g/mol) 335.407