Läs mer
Tocris Bioscience™ SB 674042
GREENER_CHOICE
Produktkod. 30002047
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30002047 50 mg
30002039 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30002047 Leverantör Tocris Bioscience™ Leverantörsnummer 4673/50
Potent and selective non-peptide OX1 antagonist
Potent and selective non-peptide orexin OX1 receptor antagonist (Kd = 3.76 nM). Exhibits 100-fold selectivity for OX1 over OX2 receptors. Has no effect at serotonergic, dopaminergic, adrenergic or purinergic receptors. Inhibits orexin 1-induced Ca2+ mobilization in CHO-DG44 cells stably transfected with the OX1 receptor.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 483313-22-0
Molekylformel C24H21FN4O2S
Specifikationer
Kemiskt namn eller material SB 674042
Kvantitet 50 mg
Formel vikt 448.51
Molekylvikt (g/mol) 448.516