Läs mer
SB-705498, MedChemExpress
Produktkod. 16450437
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16450437 50 mg
16435056 100 mg
16426787 5 mg
16404088 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16450437 Leverantör Medchem Express Leverantörsnummer HY10633
MedChemExpress SB-705498 is a potent, selective and orally bioavailable transient receptor potential vanilloid 1 (TRPV1) receptor antagonist with a pIC50 of 7.1.
SB-705498 is a potent, selective and orally bioavailable transient receptor potential vanilloid 1 (TRPV1) receptor antagonist with a pIC50 of 7.1.
Specifikationer
Kemiskt namn eller material SB-705498
CAS 501951-42-4
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C17H16BrF3N4O
Kvantitet 50 mg
Löslighetsinformation DMSO : 100 mg/mL (232.98 mM; Need ultrasonic)
LEDER BrC1=CC=CC=C1NC(N[C@@H]2CCN(C3=NC=C(C(F)(F)F)C=C3)C2)=O
Molekylvikt (g/mol) 429.23
Formel vikt 429.23
Procent renhet 98.33%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation