Läs mer
Tocris Bioscience™ SB 743921 hydrochloride
Beskrivning
SB 743921 hydrochloride is a potent kinesin spindle protein (KSP) inhibitor (Ki = 0.1 nM). Induces cell mitotic arrest and apoptosis in vitro. Inhibits the growth of a range of tumor cells in vitro, including colon (HCT 116), prostate (PC-3) and leukemia (K-562) cancer cell lines. Causes tumor regression in human tumor xenograft models in vivo, including colon (Colo205), lung (H69) and breast (MCF7) cancer cell xenografts.
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-(3-Aminopropyl)-N-[(1R)-1-[7-chloro-4-oxo-3-(phenylmethyl)-4H-1-benzopyran-2-yl]-2-methylpropyl]-4-methylbenzamide hydrochloride |
| CAS | 940929-33-9 |
| Kvantitet | 10 mg |
| Mål | Kinesin Inhibitors |
| Molekylformel | C31H33ClN2O3.HCl |
| Renhet | 98% |