Beskrivning
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifikationer
Specifikationer
| Kemiskt namn eller material | SCA-13C-15N2 hydrochloride |
| Förpackning | Glass Bottle |
| CAS | 1173020-16-0 |
| Färg | White |
| Smältpunkt | 173°C to 176°C |
| Synonym | Semicarbazide-13C-15N2hydrochloride |
| Molekylvikt (g/mol) | 114.51 |
| Kvantitet | 10 mg |
| Kvalitet | Analytical Standard |