missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAMP3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26888-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene product belongs to the SCAMP family of proteins which are secretory carrier membrane proteins. They function as carriers to the cell surface in post-golgi recycling pathways. Different family members are highly related products of distinct genes, and are usually expressed together. These findings suggest that the SCAMPs may function at the same site during vesicular transport rather than in separate pathways. Two transcript variants encoding different isoforms have been found for this gene.Specifications
SCAMP3 | |
Polyclonal | |
Unconjugated | |
SCAMP3 | |
C1orf3, PROPIN1, SCAMP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10067 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14828 | |
SCAMP3 | |
SCAMP3 Fusion Protein Ag25256 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |