missing translation for 'onlineSavingsMsg'
Learn More

SCAPER Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16994-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16876804

  • 1236.20 SEK / 20µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CCNA2/CDK2 regulatory protein that transiently maintains CCNA2 in the cytoplasm.
Specifications

Specifications

SCAPER
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BY12
KIAA1454, MSTP063, SCAPER, Zfp291, Zinc finger protein 291, ZNF291
Rabbit
IgG
20 μL
-20°C
Primary
117521, 244891, 49855
Immunohistochemistry (Paraffin)
Unconjugated
SCAPER
Liquid
SCAPER
SCAPER Fusion Protein Ag10679
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers