missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAPER Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SCAPER |
---|---|
Concentration | 0.2 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16876804
|
Proteintech
16994-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16866804
|
Proteintech
16994-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CCNA2/CDK2 regulatory protein that transiently maintains CCNA2 in the cytoplasm.Specifications
SCAPER | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q9BY12 | |
117521, 244891, 49855 | |
SCAPER Fusion Protein Ag10679 | |
Primary | |
-20°C | |
SCAPER |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
KIAA1454, MSTP063, SCAPER, Zfp291, Zinc finger protein 291, ZNF291 | |
SCAPER | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |