Läs mer
SCH 563705, MedChemExpress
Produktkod. 16432778
Encompass_Preferred
Change view
Kvantitet:
2 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16432778 10 mg
16469995 2 mg
16425517 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16432778 Leverantör Medchem Express Leverantörsnummer HY10011
MedChemExpress SCH 563705 is a potent and orally available CXCR2 and CXCR1 antagonist, with IC50s of 1.3 nM, 7.3 nM and Kis of 1 and 3 nM, respectively.
SCH 563705 is a potent and orally available CXCR2 and CXCR1 antagonist, with IC50s of 1.3 nM, 7.3 nM and Kis of 1 and 3 nM, respectively.
Specifikationer
Kemiskt namn eller material SCH 563705
CAS 473728-58-4
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C23H27N3O5
Kvantitet 10 mg
Löslighetsinformation DMSO : ≥ 30 mg/mL (70.51 mM)
LEDER O=C(C(NC1=CC=CC(C(N(C)C)=O)=C1O)=C2N[C@H](CC)C3=CC(C(C)C)=CO3)C2=O
Molekylvikt (g/mol) 425.48
Formel vikt 425.48
Procent renhet 98.2%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation