Läs mer
Tocris Bioscience™ SCH 79797 dihydrochloride
GREENER_CHOICE
Produktkod. 18086052
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18086052 10 mg
30001269 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18086052 Leverantör Tocris Bioscience™ Leverantörsnummer 1592/10
Potent, selective non-peptide PAR1 antagonist
Potent, selective non-peptide PAR1 receptor antagonist (IC50 = 70 nM). Inhibits haTRAP-induced- but not γ-thrombin-, ADP- or collagen-induced human platelet aggregation. Also selectively blocks PAR1 agonist- or thrombin-induced increases in cytosolic Ca2+ in vascular smooth muscle cells.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1216720-69-2
Molekylformel C23H27Cl2N5
Specifikationer
Kemiskt namn eller material SCH 79797 dihydrochloride
Kvantitet 10 mg
Formel vikt 444.41
Renhet >99%
Molekylvikt (g/mol) 444.404