Läs mer
SCH00013, MedChemExpress
Produktkod. 16445867
Encompass_Preferred
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16445867 5 mg
16430689 1 mg
16403138 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16445867 Leverantör Medchem Express Leverantörsnummer HY100718
MedChemExpress SCH00013 is a cardiotonic agent that primarily acts via an increase in myofibrillar Ca++ sensitivity, have a significant Ca(2+)sensitizing effect at pH 7.2 to 7.4.
SCH00013 is a cardiotonic agent that primarily acts via an increase in myofibrillar Ca++ sensitivity, have a significant Ca(2+)sensitizing effect at pH 7.2 to 7.4.
Specifikationer
Kemiskt namn eller material SCH00013
Fysisk form Solid
CAS 217963-18-3
Färg Light Yellow
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C18H20N4O2
För användning med (applikation) Neuroscience-Neuromodulation
LEDER N#CC1=CC=C(C(O)CN2CC=C(C(CC3)=NNC3=O)CC2)C=C1
Molekylvikt (g/mol) 324.38
Formel vikt 324.38
Procent renhet 98.04%