Läs mer
Schisantherin A, MedChemExpress
Produktkod. 16469068
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16469068 10 mg
16414877 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16469068 Leverantör Medchem Express Leverantörsnummer HYN0694
MedChemExpress Schisantherin A is a dibenzocyclooctadiene lignan. Schisantherin A inhibits p65-NF-κB translocation into the nucleus by IκBα degradation.
Schisantherin A is a dibenzocyclooctadiene lignan. Schisantherin A inhibits p65-NF-κB translocation into the nucleus by IκBα degradation.
Specifikationer
Kemiskt namn eller material Schisantherin A
CAS 58546-56-8
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C30H32O9
Kvantitet 10 mg
Synonym Gomisin-C Schizantherin-A Wuweizi ester-A
Löslighetsinformation DMSO : ≥ 100 mg/mL (186.37 mM)
LEDER O[C@]([C@@H](C)C1)(C)[C@@H](OC(C2=CC=CC=C2)=O)C3=CC(OC)=C(OC)C(OC)=C3C4=C1C=C5OCOC5=C4OC
Molekylvikt (g/mol) 536.57
Formel vikt 536.57
Procent renhet 99.43%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation