Läs mer
SI Analytics™ ScienceLine Ion Selective Electrode
Produktkod. 10516063 Handla allt SI Analytics Produkter
Change view
Parameter:
Bromid
Jodid
Kalium
Klorid
Koppar
Sulfid
pH-intervall:
0 till 14
1 till 12
2 till 12
2 till 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Parameter pH-intervall
10516063 Jodid 0 till 14
10129532 Klorid 2 till 12
10405063 Bromid 1 till 12
10405253 Koppar 2 till 6
10765493 Kalium 2 till 12
10211503 Sulfid 2 till 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Produktkod. 10516063 Leverantör SI Analytics™ Leverantörsnummer 285130410
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
SI Analytics ScienceLine Ion Selective Electrodes possess the advantage of a much lower temperature sensitivity and a metal ion free reference system.
  • ScienceLine sensors for ammonia, sodium, oxygen, ion-selective indicator electrodes
  • Also measure pH and temperature
TRUSTED_SUSTAINABILITY
Specifikationer
pH-intervall 0 till 14
Modell I 60
Elektrod Selective
Temperaturområde (metriskt). 0°C to 80°C
Typ Ion Selective Electrode
Mätområde 0.006 to 127,000 mg/L
Parameter Jodid