missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Beskrivning
Immunogen sequence: EHLYVNTQGL DAPEPEDSPK KDLFDMRPFE DALKLHECSV AAGVTAAPLP LEDQWPSPPT RRAPVAPTEE QLRQEPWYHG RMSRRAAERM LRADGD Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 85%.
Src homology 2 (SH2) domains bind specifically to tyrosine-phosphorylated proteins that temporally participate in signal transduction events. Shc-like protein (Sck) is a neuronal adaptor protein that contains an N-terminal PTB (phosphotyrosine binding) domain, a collagen homology (CH) domain, and a conserved C-terminal SH2 domain. Human Sck transcripts are present at high levels in liver, pancreas, prostate and ovary. In vascular endothelial cells, Sck participates in VEGF-induced signal transduction. Treatment of human umbilical vein endothelial (HUVEC) cells with VEGF induces recruitment of Sck to tyrosine-1175 of the kinase insert domain-containing receptor (KDR) and enhances Sck tyrosine phosphorylation.
Specifikationer
Specifikationer
| Antigen | SCK |
| Användningsområden | Immunohistochemistry (Paraffin), Western Blot |
| Klassificering | Polyclonal |
| Koncentration | 0.1 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gen | SHC2 |
| Genaccessionsnr. | P98077 |
| Gene Alias | 6720466E06; neuronal Shc adaptor homolog; protein Sck; Protein Sli; SCK; SH2 domain protein C2; SHC (Src homology 2 domain containing) transforming protein 2; SHC adaptor protein 2; SHC2; ShcB; SHC-transforming protein 2; SHC-transforming protein B; SLI; src homology 2 domain-containing transforming protein C2; src homology 2 domain-containing-transforming protein C2 |
| Gensymboler | SHC2 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?
