missing translation for 'onlineSavingsMsg'
Learn More

SCNN1B Recombinant Protein, Abnova

Human SCNN1B full-length ORF (AAH36352, 1 a.a. - 640 a.a.) recombinant protein with GST-tag at N-terminal

Brand:  Abnova H00006338-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16180542

  • 5030.41 SEK / 25µg

Estimated Shipment: 03-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Nonvoltage-gated, amiloride-sensitive, sodium channels control fluid and electrolyte transport across epithelia in many organs. These channels are heteromeric complexes consisting of 3 subunits: alpha, beta, and gamma. This gene encodes the beta subunit.

  • Molecular weight: 96.14kDa
  • Preparation method: in vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 Fast Flow
  • Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH 8 in the elution buffer
  • Quality Control Testing: 12.5% SDS-PAGE stained with Coomassie Blue

Sequence:
MHVKKYLLKGLHRLQKGPGYTYKELLVWYCDNTNTHGPKRTICEGPKKKAMWFLLTLLFAALVCWQWGIFIRTYLSWEVSVSLSVGFKTMDFPAVTICNASPFKYSKIKHLLKDLDELMEAVLERILAPELSHANATRNLNFSIWNHTPLVLIDERNPHHPMVLDLFGDNHNGLTSSSASEKICNAHGCKMAMRLCSLNRTQCTFRNFTSATQALTEWYILQATNIFAQVPQQELVEMSYPGEQMILACLFGAEPCNYRNFTSIFYPHYGNCYIFNWGMTEKALPSANPGTEFGLKLILDIGQEDYVPFLASTAGVRLMLHEQRSYPFIRDEGIYAMSGTETSIGVLVDKLQRMGEPYSPCTVNGSEVPVQNFYSDYNTTYSIQACLRSCFQDHMIRNCNCGHYLYPLPRGEKYCNNRDFPDWAHCYSDLQMSVAQRETCIGMCKESCNDTQYKMTISMADWPSEASEDWIFHVLSQERDQSTNITLSRKGIVKLNIYFQEFNYRTIEESAANNIVWLLSNLGGQFGFWMGGSVLCLIEFGEIIIDFVWITIIKLVALAKSLRQRRAQASYAGPPPTVAELVEAHTNFGFQPDTAPRSPNTGPYPSEQALPIPGTPPPNYDSLRLQPLDVIESDSEGDAI

Best use within three months from the date of receipt of this protein.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH36352
Solution
6338
SCNN1B (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SCNN1B
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
96.14
8
Glutathione Sepharose 4 Fast Flow
25 ug
MHVKKYLLKGLHRLQKGPGYTYKELLVWYCDNTNTHGPKRTICEGPKKKAMWFLLTLLFAALVCWQWGIFIRTYLSWEVSVSLSVGFKTMDFPAVTICNASPFKYSKIKHLLKDLDELMEAVLERILAPELSHANATRNLNFSIWNHTPLVLIDERNPHHPMVLDLFGDNHNGLTSSSASEKICNAHGCKMAMRLCSLNRTQCTFRNFTSATQALTEWYILQATNIFAQVPQQELVEMSYPGEQMILACLFGAEPCNYRNFTSIFYPHYGNCYIFNWGMTEKALPSANPGTEFGLKLILDIGQEDYVPFLASTAGVRLMLHEQRSYPFIRDEGIYAMSGTETSIGVLVDKLQRMGEPYSPCTVNGSEVPVQNFYSDYNTTYSIQACLRSCFQDHMIRNCNCGHYLYPLPRGEKYCNNRDFPDWAHCYSDLQMSVAQRETCIGMCKESCNDTQYKMTISMADWPSEASEDWIFHVLSQERDQSTNITLSRKGIVKLNIYFQEFNYRTIEESAANNIVWLLSNLGGQFGFWMGGSVLCLIEFGEIIIDFVWITIIKLVALAKSLRQRRAQASYAGPPPTVAELVEAHTNFGFQPDTAPRSPNTGPYPSEQALPIPGTPPPNYDSLRLQPLDVIESDSEGDAI
ENaCb/ENaCbeta/SCNEB
SCNN1B
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates