Läs mer
Scopine Di(2-thienylglycolate), TRC
Produktkod. 30426426
Change view
Kvantitet:
10 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30426426 10 mg
30415769 1 g
30417588 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30426426 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS19996510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 136310-64-0
Molekylformel C18 H19 N O4 S2
Molekylvikt (g/mol) 377.48
Synonym 2-Thiopheneacetic acid, α-hydroxy-α-2-thienyl-, 9-methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl ester, (1α,2β,4β,5α,7β)-, (1α,2β,4β,5α,7β)-7-[(Hydroxydi(2-thienyl)acetyl)oxy]-9-methyl-3-oxa-9-azatricyclo[3.3.1.0]nonane, Di(2-thienyl)glycolic acid scopine ester, Dithienylglycolic acid scopine ester, N-Demethyl tiotropium, Scopine di(2-thienylglycolate), Scopine hydroxydi(2-thienyl)acetate, 2-Thiopheneacetic acid, α-hydroxy-α-2-thienyl-, (1α,2β,4β,5α,7β)-9-methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl ester, Tiotropium Bromide Monohydrate Imp B (EP)
IUPAC-namn (1R,2R,4S,5S,7s)-9-Methyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-7-yl 2-Hydroxy-2,2-dithiophen-2-ylacetate
LEDER CN1[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)C(O)(c4cccs4)c5cccs5
Specifikationer
Kemiskt namn eller material (1R,2R,4S,5S,7s)-9-Methyl-3-oxa-9-azatricyclo(3.3.1.02,4)nonan-7-yl 2-Hydroxy-2,2-dithiophen-2-ylacetate
Kvantitet 10 mg
InChI formel InChI=1S/C18H19NO4S2/c1-19-11-8-10(9-12(19)16-15(11)23-16)22-17(20)18(21,13-4-2-6-24-13)14-5-3-7-25-14/h2-7,10-12,15-16,21H,8-9H2,1H3/t10-,11-,12+,15-,16+
Formel vikt 377.0755