Läs mer
Scopine Methiodide, TRC
Produktkod. 30427291
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30427291 10 mg
1 options
Produktkod. 30427291 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS20003510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Scopine Methiodide
CAS 21662-36-2
Molekylformel C9H16INO2
Kvantitet 10 mg
Synonym (1R,2R,4S,5S,7s)-7-Hydroxy-9,9-dimethyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-9-ium Iodide, N-Methylscopine Iodide, Methscopinium Iodide, 3-Oxa-9-azatricyclo[3.3.1.02,4]nonane 3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane Derivative, 6beta,7beta-Epoxy-3alpha-hydroxy-8-methyl-1alphaH,5alphaH-tropanium Iodide, 1alpha,2beta,4beta,5alpha,7beta)-7-Hydroxy-9,9-dimethyl-3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane Iodide
InChI formel InChI=1S/C9H16NO2.HI/c1-10(2)6-3-5(11)4-7(10)9-8(6)12-9;/h5-9,11H,3-4H2,1-2H3;1H/q+1;/p-1/t5-,6-,7+,8-,9+;
LEDER [I-].C[N+]1(C)[C@@H]2C[C@@H](O)C[C@H]1[C@@H]3O[C@H]23
Molekylvikt (g/mol) 297.13
Formel vikt 297.0226