Läs mer
Scopine Methobromide, TRC
Produktkod. 17921845
Change view
Kvantitet:
2.5 mg
10 mg
100 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17921845 2.5 mg 2.5mg
17902055 100 mg -
17904285 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17921845 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS1999852.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1508-46-9
Molekylformel C9 H16 N O2 . Br
Molekylvikt (g/mol) 250.13
Synonym (1R,2R,4S,5S,7s)-7-Hydroxy-9,9-dimethyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane Bromide, 1αH,5αH-Tropanium, 6β,7β-epoxy-3α-hydroxy-8-methyl-, bromide (8CI), 3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane, 7-hydroxy-9,9-dimethyl-, bromide, (1α,2β,4β,5α,7β)- (9CI), N-Methylscopinium bromide (7CI), 3-Oxa-9-azatricyclo[3.3.1.02,4]nonane, 3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane deriv., BEA 2180 alcohol, Methscopinium bromide, N-Methylscopine bromide, Scopine methobromide, 3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane, 7-hydroxy-9,9-dimethyl-, bromide (1:1), (1R,2R,4S,5S)-rel-, Tiotropium Bromide Monohydrate Imp. G (EP), (1R,2R,4S,5S,7s)-7-Hydroxy-9,9-dimethyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-9-ium Bromide
LEDER [Br-].C[N+]1(C)[C@@H]2C[C@@H](O)C[C@H]1[C@@H]3O[C@H]23
Specifikationer
Kemiskt namn eller material Scopine Methobromide
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C9H16NO2.BrH/c1-10(2)6-3-5(11)4-7(10)9-8(6)12-9;/h5-9,11H,3-4H2,1-2H3;1H/q+1;/p-1/t5-,6-,7+,8-,9+;
Formel vikt 249.0364
Procent renhet >95%