Läs mer
Scopine, Mikromol™
Produktkod. 30051761
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051761 25 mg
30050065 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051761 Leverantör Mikromol Leverantörsnummer MM0546.090025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Scopine
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Scopine
CAS 498-45-3
Analyt- eller komponentnamn Scopine
Frakt skick Room Temperature
Molekylformel C8 H13 N O2
Certifikat och efterlevnad ISO 9001
Synonym (1α,2β,4β,5α,7β)-9-methyl-3-Oxa-9-azatricyclo[3.3.1.02,4]nonan-7-ol, 1αH,5αH-Tropan-3α-ol, 6β,7β-epoxy- (8CI), Scopine (6CI,7CI), (1α,2β,4β,5α,7β)-9-Methyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-7-ol, 6,7-Epoxytropine, Scopanol, Scopin
InChI formel InChI=1S/C8H13NO2/c1-9-5-2-4(10)3-6(9)8-7(5)11-8/h4-8,10H,2-3H2,1H3/t4-,5-,6+,7-,8+
LEDER CN1[C@@H]2C[C@@H](O)C[C@H]1[C@@H]3O[C@H]23
IUPAC-namn (1α,2β,4β,5α,7β)-9-Methyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-7-ol
Molekylvikt (g/mol) 155.19
Kvantitet 25 mg
Formel vikt 155.0946 g/mol