Läs mer
Scopolamine 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifikationer
Specifikationer
| Kemiskt namn eller material | Hyoscine |
| CAS | 51-34-3 |
| Analyt- eller komponentnamn | Hyoscine |
| Frakt skick | +20°C |
| Molekylformel | C17 H21 N O4 |
| Koncentration | 100 μg/mL |
| Certifikat och efterlevnad | ISO 17034 |
| Synonym | Hyoscine, (1R,2R,4S,5S,7s)-9-Methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl (2S)-3-hydroxy-2-phenylpropanoate, Benzeneacetic acid, α-(hydroxymethyl)-, (1α,2β,4β,5α,7β)-9-methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl ester, (αS)-, 1αH,5αH-Tropan-3α-ol, 6β,7β-epoxy-, (-)-tropate (ester) (8CI), Benzeneacetic acid, α-(hydroxymethyl)-, 9-methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl ester, [7(S)-(1α,2β,4β,5α,7β)]-, 3-Oxa-9-azatricyclo[3.3.1.02,4]nonane, benzeneacetic acid deriv., (-)-Hyoscine, (-)-Scopolamine, 6,7-Epoxytropine tropate, 6β,7β-Epoxy-3α-tropanyl S-(-)-tropate, 9-Methyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-7-ol (-)-tropate, Atrochin, Atroquin, SEE, Scop, Scopine (-)-tropate, Scopine tropate, Scopoderm TTS, Scopolamine, Transcop, Transderm-Scop, Tropic acid ester with scopine, l-Scopolamine |
| InChI formel | InChI=1S/C17H21NO4/c1-18-13-7-11(8-14(18)16-15(13)22-16)21-17(20)12(9-19)10-5-3-2-4-6-10/h2-6,11-16,19H,7-9H2,1H3/t11-,12-,13-,14+,15-,16+/m1/s1 |
| LEDER | CN1[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)[C@H](CO)c4ccccc4 |
| Visa mer |