Läs mer
Scopolamine Beta-D-Glucuronide, TRC
Produktkod. 30136460
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136460 10 mg
30005190 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136460 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS20001010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17660-02-5
Molekylformel C23 H29 N O10
Molekylvikt (g/mol) 479.48
Synonym Scopolamine-B-D-Glucuronide
LEDER CN1[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)[C@H](CO[C@@H]4O[C@@H]([C@@H](O)[C@H](O)[C@H]4O)C(=O)O)c5ccccc5
Specifikationer
Kemiskt namn eller material Scopolamine beta-D-Glucuronide
Kvantitet 10 mg
InChI formel InChI=1S/C23H29NO10/c1-24-13-7-11(8-14(24)19-18(13)33-19)32-22(30)12(10-5-3-2-4-6-10)9-31-23-17(27)15(25)16(26)20(34-23)21(28)29/h2-6,11-20,23,25-27H,7-9H2,1H3,(H,28,29)/t11-,12-,13-,14+,15+,16+,17-,18-,19+,20+,23-/m1/s1
Formel vikt 479.1791