Läs mer
Scopolamine Methyl Bromide, TRC
Produktkod. 30004954
Change view
Kvantitet:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004954 5 g
30004351 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30004954 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS2000505G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 155-41-9
Molekylformel C18 H24 N O4 . Br
Molekylvikt (g/mol) 398.29
Synonym (1R,2R,4S,5S,7s)-7-[[(2S)-3-Hydroxy-2-phenylpropanoyl]oxy]-9,9-dimethyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane bromide, Methylhyoscine bromide, 3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane, 7-[(2S)-3-hydroxy-1-oxo-2-phenylpropoxy]-9,9-dimethyl-, bromide (1:1), (1α,2β,4β,5α,7β)-, 1αH,5αH-Tropanium, 6β,7β-epoxy-3α-hydroxy-8-methyl-, bromide, (-)-tropate (8CI), 3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane, 7-(3-hydroxy-1-oxo-2-phenylpropoxy)-9,9-dimethyl-, bromide, [7(S)-(1α,2β,4β,5α,7β)]-, 3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane, 7-[(2S)-3-hydroxy-1-oxo-2-phenylpropoxy]-9,9-dimethyl-, bromide, (1α,2β,4β,5α,7β)- (9CI), N-Methylscopolammonium bromide (6CI,7CI), 3-Oxa-9-azatricyclo[3.3.1.02,4]nonane, 3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane deriv., Tropic acid, (-), ester with 6β,7β-epoxy-3α-hydroxy-8-methyl-1αH,5αH-tropanium bromide (8CI), (-)-Scopolamine methobromide, (-)-Scopolamine methyl bromide, Ampyrox, Blocan, Diopal, Epoxymethamine bromide, Epoxytropine tropate methylbromide, Holopon, Hyoscine methobromide, Hyoscine methyl bromide, Lescopine bromide, Mescopil, Methscopolamin bromide, Methscopolamine bromide, Methylscopolamine bromide, N-Methylscopolamine bromide, N-Methylscopolaminium bromide, NSC 120606, NSC 61809, Neo-Avagal, Nutrop, Pamine, Pamine bromide, Paraspan, Proscomide, Restropin, Scopolamine methobromide, Scopolamine methyl bromide, U 5036
IUPAC-namn (1R,2R,4S,5S,7s)-7-[[(2S)-3-Hydroxy-2-phenylpropanoyl]oxy]-9,9-dimethyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane bromide
LEDER [Br-].C[N+]1(C)[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)[C@H](CO)c4ccccc4
Specifikationer
Kemiskt namn eller material Methscopolamine Bromide
Kvantitet 5 g
InChI formel InChI=1S/C18H24NO4.BrH/c1-19(2)14-8-12(9-15(19)17-16(14)23-17)22-18(21)13(10-20)11-6-4-3-5-7-11;/h3-7,12-17,20H,8-10H2,1-2H3;1H/q+1;/p-1/t12-,13-,14-,15+,16-,17+;/m1./s1
Formel vikt 397.0889