Läs mer
Scopolamine N-Oxide, TRC
Produktkod. 30134289
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134289 100 mg
30005098 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134289 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS200020100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 97-75-6
Molekylformel C17H21NO5
Molekylvikt (g/mol) 319.35
Synonym (αS)-α-(Hydroxymethyl)benzeneacetic Acid (1α,2β,4β,5α,7β)-9-Methyl-9-oxido-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl Ester, 6β,7β-Epoxy-1αH,5αH-tropan-3α-ol (-)-Tropate 8-Oxide, [7(S)-(1α,2β,4β,5α,7β)]-α-(Hydroxymethyl)benzeneacetic Acid 9-Methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl Ester N-Oxide, Scopolamine, N-Oxide, 3-Oxa-9-azatricyclo[3.3.1.02,4]nonane Benzeneacetic Acid Deriv., Genoscopolamine, Hyoscinamine Oxide, Hyoscine N-Oxide, Hyoscineamine Oxide, Oxicopolamina, Scolamine, Scolen, Scopodex, Scopolamine aminoxide, Seamalin, Seronex
IUPAC-namn [(1R,2R,4S,5S)-9-methyl-9-oxido-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonan-7-yl] (2S)-3-hydroxy-2-phenylpropanoate
LEDER O=C([C@H](CO)C1=CC=CC=C1)O[C@H]2C[C@H]3[C@H](O4)[C@H]4[C@@H](C2)N3(C)=O
Specifikationer
Kemiskt namn eller material Scopolamine N-Oxide
Kvantitet 100 mg
InChI formel InChI=1S/C17H21NO5/c1-18(21)13-7-11(8-14(18)16-15(13)23-16)22-17(20)12(9-19)10-5-3-2-4-6-10/h2-6,11-16,19H,7-9H2,1H3/t11-,12-,13-,14+,15-,16+,18+/m1/s1
Formel vikt 319.142