Läs mer
Tocris Bioscience™ SCOTfluor 510 Fmoc-Dapa-OH
Beskrivning
SCOTfluor 510 Fmoc-Dapa-OH is a Fmoc protected fluorescent amino acid. Used as a building block in solid-phase peptide synthesis to prepare PAINT imaging probes. Application: use in solid-phase peptide synthesis (SPPS) for PAINT imaging. Properties and Photophysical Data: excitation and emission maxima (λ) are 488 nm and 601 nm, respectively; quantum yield = 0.19; extinction coefficient = 4,480 M-1cm-1.
Specifikationer
Specifikationer
| Kemiskt namn eller material | (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-((7-nitrobenzo[c][1,2,5]selenadiazol-4-yl)amino) propanoic acid |
| CAS | 3023933-33-4 |
| Kvantitet | 25 mg |
| Mål | Fluorescent Probes |
| Molekylformel | C24H19N5O6Se |
| Renhet | 0.95 |