Learn More
DWK Life Sciences Screw Cap, for Culture tubes, with seal
This screw cap is used to close culture tubes with threads (threads not according to DIN).
269.00 SEK
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Polypropylene, TPE |
For Use With (Application) | Growth of microorganisms; Storage and transport of small quantities of liquid; Working with cell culture media under sterile conditions in biosafety cabinets and clean hoods |
Quantity | 100/Pk. |
Product Code | Brand | Closure Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Price | Quantity & Availability | |||||
10521275
|
DWK Life Sciences
299901307 |
GL18 |
269.00 SEK
Pack of 100 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
10090414
|
DWK Life Sciences
299901204 |
GL14 |
269.00 SEK
Pack of 100 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
The in vitro cultivation of animal and plant cells in a nutrient medium is an essential part of today's research in the field of microbiology. These cultivation approaches can take place at different scales - the vessels are selected accordingly. For test runs, rather small vessels such as culture tubes are used. In addition to culture tubes made of DURAN™, our product range also includes culture tubes made of soda-lime glass. This is a glass of the third water resistance Class And belongs to the category of soda-lime glasses with a high proportion of alkali and alkaline earth oxides.
- Cultivating, storage and transport of microorganisms
Specifications
Autoclavable | |
Polypropylene, TPE | |
Growth of microorganisms; Storage and transport of small quantities of liquid; Working with cell culture media under sterile conditions in biosafety cabinets and clean hoods | |
100/Pk. |