missing translation for 'onlineSavingsMsg'
Learn More

SCYL3 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11249-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16878193

  • 4721.67 SEK / 150µL
Estimated Shipment: 02-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

May play a role in regulating cell adhesion/migration complexes in migrating cells.
Specifications

Specifications

SCYL3
Polyclonal
Unconjugated
SCYL3
Ezrin binding protein PACE 1, PACE 1, PACE1, RP1 97P20.2, SCY1 like 3 (S. cerevisiae), SCY1 like protein 3, SCYL3
Rabbit
Antigen Affinity Chromatography
RUO
57147
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8IZE3
SCYL3
SCYL3 Fusion Protein Ag1769
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers