missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCYL3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11249-1-AP-20UL
Informations supplémentaires : Weight : 0.00010kg
Description
May play a role in regulating cell adhesion/migration complexes in migrating cells.Spécification
SCYL3 | |
Polyclonal | |
Unconjugated | |
SCYL3 | |
Ezrin binding protein PACE 1, PACE 1, PACE1, RP1 97P20.2, SCY1 like 3 (S. cerevisiae), SCY1 like protein 3, SCYL3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57147 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8IZE3 | |
SCYL3 | |
SCYL3 Fusion Protein Ag1769 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |