missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDHC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14575-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes one of four nuclear-encoded subunits that comprise succinate dehydrogenase, also known as mitochondrial complex II, a key enzyme complex of the tricarboxylic acid cycle and aerobic respiratory chains of mitochondria. The encoded protein is one of two integral membrane proteins that anchor other subunits of the complex, which form the catalytic core, to the inner mitochondrial membrane. There are several related pseudogenes for this gene on different chromosomes. Mutations in this gene have been associated with paragangliomas. Alternatively spliced transcript variants have been described.Specifications
SDHC | |
Polyclonal | |
Unconjugated | |
SDHC | |
CYB560, CYBL, PGL3, QPs 1, QPS1, SDH3, SDHC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
289217, 6391, 66052 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99643, Q9CZB0 | |
Sdhc | |
SDHC Fusion Protein Ag5775 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |