Learn More
SDPR Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12339-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16840004
  • 3313.85 SEK / 150µL
Estimated Shipment: 03-10-2022
to see stock.
Description
Description
The SDPR antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SDPR. This antibody recognizes human, mouse, rat antigen. The SDPR antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Specifications
Specifications
SDPR
Polyclonal
Unconjugated
150 μL
8436
IgG
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
333 μg/mL
Rabbit
Primary
Mouse, Rat, Human