missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ SDS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558704
This item is not returnable.
View return policy
Description
Immunogen sequence: PATIVVPSTT PALTIERLKN EGATVKVVGE LLDEAFELAK ALAKNNPGWV YIPPFDDPLI WEGHASIVKE LKE Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 89%.
SDS encodes one of three enzymes that are involved in metabolizing serine and glycine. L-serine dehydratase converts L-serine to pyruvate and ammonia and requires pyridoxal phosphate as a cofactor. The encoded protein can also metabolize threonine to NH4+ and 2-ketobutyrate. The encoded protein is found predominantly in the liver.
Specifications
| SDS | |
| Polyclonal | |
| Unconjugated | |
| SDS | |
| 4432411H13Rik; L-serine ammonia-lyase; L-serine deaminase; L-serine dehydratase; L-serine dehydratase/L-threonine deaminase; L-threonine dehydratase; RATSDHE1; Sdh; SDH2; Sdhe1; SDS; serine dehydratase; TDH | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10993 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P20132 | |
| SDS | |
| Recombinant protein corresponding to Human SDS. Recombinant protein control fragment (Product #RP-97731). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction