missing translation for 'onlineSavingsMsg'
Learn More

SDSL Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16242-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16825814

  • 3313.85 SEK / 150µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SDSL has low serine dehydratase and threonine dehydratase activity.
Specifications

Specifications

SDSL
0.36 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8R238, Q96GA7
L serine deaminase, L threonine dehydratase, SDH 2, SDS RS1, SDSL, Serine dehydratase 2, serine dehydratase like, TDH
Rabbit
IgG
150 μL
-20°C
Primary
113675, 257635
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SDSL
Liquid
SDSL
SDSL Fusion Protein Ag9114
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documents
Certificates
Special Offers

Special Offers