Läs mer
SDZ 285-428, TRC
Produktkod. 30412218
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30412218 5 mg
30420570 10 mg
30418695 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30412218 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS2111135MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 174262-13-6
Molekylformel C24H20ClN3O
Molekylvikt (g/mol) 401.89
Synonym SDZ 285428, 4'-Chloro-N-[2-(1H-imidazol-1-yl)-2-phenylethyl][1,1'-biphenyl]-4-carboxamide
IUPAC-namn N-(2-(1H-imidazol-1-yl)-2-phenylethyl)-4'-chloro-[1,1'-biphenyl]-4-carboxamide
LEDER O=C(C1=CC=C(C2=CC=C(Cl)C=C2)C=C1)NCC(N3C=NC=C3)C4=CC=CC=C4
Specifikationer
Kemiskt namn eller material 4'-Chloro-N-[2-(1H-imidazol-1-yl)-2-phenylethyl][1,1'-biphenyl]-4-carboxamide
Kvantitet 5 mg
InChI formel InChI=1S/C24H20ClN3O/c25-22-12-10-19(11-13-22)18-6-8-21(9-7-18)24(29)27-16-23(28-15-14-26-17-28)20-4-2-1-3-5-20/h1-15,17,23H,16H2,(H,27,29)
Formel vikt 401.13