Läs mer
Tocris Bioscience™ SDZ NKT 343
Beskrivning
Highly selective human tachykinin NK1 receptor antagonist (IC50 values are 0.62 and 451 nM for human and rat receptors respectively) that displays > 130-fold selectivity over human NK2 and NK3 receptors. Potently antagonizes SP-induced Ca2+ efflux in vitro and inhibits mechanical hyperalgesia in vivo.
Specifikationer
Specifikationer
| CAS | 180046-99-5 |
| Kvantitet | 10 mg |
| Produkttyp | SDZ NKT 343 |
| Renhet | >98% |
| Molekylvikt (g/mol) | 579.657 |