Läs mer
SE 563, TRC
Produktkod. 30414151
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30414151 10 mg
30428415 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30414151 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS21130010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 174819-21-7
Molekylformel C22 H17 Cl F3 N O3
Molekylvikt (g/mol) 435.82
Synonym 2H-3,1-Benzoxazin-2-one, 6-chloro-4-(2-cyclopropylethynyl)-1,4-dihydro-1-[(4-methoxyphenyl)methyl]-4-(trifluoromethyl)-, (4S)-, 2H-3,1-Benzoxazin-2-one, 6-chloro-4-(cyclopropylethynyl)-1,4-dihydro-1-[(4-methoxyphenyl)methyl]-4-(trifluoromethyl)-, (4S)- (9CI), 2H-3,1-Benzoxazin-2-one, 6-chloro-4-(cyclopropylethynyl)-1,4-dihydro-1-[(4-methoxyphenyl)methyl]-4-(trifluoromethyl)-, (S)-, SE 563
IUPAC-namn (4S)-6-chloro-4-(2-cyclopropylethynyl)-1-[(4-methoxyphenyl)methyl]-4-(trifluoromethyl)-3,1-benzoxazin-2-one
LEDER COc1ccc(CN2C(=O)O[C@@](C#CC3CC3)(c4cc(Cl)ccc24)C(F)(F)F)cc1
Specifikationer
Kemiskt namn eller material SE 563
Kvantitet 10 mg
InChI formel InChI=1S/C22H17ClF3NO3/c1-29-17-7-4-15(5-8-17)13-27-19-9-6-16(23)12-18(19)21(22(24,25)26,30-20(27)28)11-10-14-2-3-14/h4-9,12,14H,2-3,13H2,1H3/t21-/m0/s1
Formel vikt 435.0849