Learn More
Seal (O-ring 56 x 3.6 mm), Supelco™
Product Code. 15346498
Change view
Click to view available options
Quantity:
1 U
Unit Size:
Each
Product Code. Quantity Unit Size
15346498 1 U Each
1 options
Product Code. 15346498 Supplier Supelco Supplier No. 9671001047
Beräknad leverans: 25-11-2026
COUPLING VA AM1/2NF 1/2 in. 1 Pc.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Withdrawl Systems (Products 1.01114 and 1.01123)
Quantity 1 U