Learn More
Seasonal H1N1 Neuraminidase Antibody (10C5F12), HRP, R&D Systems™
Product Code. 18601347 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity Unit Size
18601347 0.1 mL 0.1mL
1 options
Product Code. 18601347 Supplier Novus Biologicals Supplier No. NBP241331H
 Restricted Item
Beräknad leverans: 04-12-2026
A Novus product, part of R&D Systems. Mouse Monoclonal Antibody
Seasonal H1N1 Neuraminidase Monoclonal antibody specifically detects Seasonal H1N1 Neuraminidase in Influenza Virus N1, Virus samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Antigen Seasonal H1N1 Neuraminidase
Applications ELISA
Classification Monoclonal
Clone 10C5F12
Conjugate HRP
Formulation PBS
Gene Alias EC=3.2.1.18, Neuraminidase
Host Species Mouse
Immunogen Antibody was raised against a synthetic peptide from the seasonal influenza (H1N1) neuraminidase protein.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Target Species Influenza virus N1, Virus
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b
Product Line Novus