Läs mer
Sec-Butyl 4-Methylbenzenesulfonate (>80%, with an impurity of pTSA), TRC
Produktkod. 30136678
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136678 100 mg
30133111 500 mg
30008906 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136678 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS282080100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 715-11-7
Molekylformel C11 H16 O3 S
Molekylvikt (g/mol) 228.31
Synonym p-Toluenesulfonic acid, sec-butyl ester (7CI,8CI), sec-Butyl alcohol, p-toluenesulfonate (6CI), 1-Methylpropyl p-toluenesulfonate
IUPAC-namn sec-butyl 4-methylbenzenesulfonate
LEDER CCC(C)OS(=O)(=O)c1ccc(C)cc1
Specifikationer
Kemiskt namn eller material sec-Butyl 4-Methylbenzenesulfonate
Kvantitet 100 mg
InChI formel InChI=1S/C11H16O3S/c1-4-10(3)14-15(12,13)11-7-5-9(2)6-8-11/h5-8,10H,4H2,1-3H3
Formel vikt 228.082